Diisopropylcarbodiimide ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC

D0254-25G D0254-250G
TCIAD0254-25GEA 855 SEK
TCIAD0254-25G TCIAD0254-250G
Diisopropylcarbodiimide ≥98.0%
Diisopropylcarbodiimide
Formel: C₇H₁₄N₂
MW: 126,2 g/mol
Kokpunkt: 145 °C (1013 hPa)
Densitet: 0,81 g/cm³ (20 °C)
Flampunkt: 31°C
MDL Number: MFCD00065689
CAS nummer: 693-13-0
EINECS: 211-743-7
UN: 2929
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b68242869f5fbdf89f99f8d17fefdf9e09fc7c8a&vid=b68242869f5fbdf89f99f8d17fefdf9e09fc7c8a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR