3-Amino-2-naphthoic acid ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
A2258-1G
A2258-5G
TCIAA2258-1GEA
495
SEK
TCIAA2258-1G
TCIAA2258-5G
3-Amino-2-naphthoic acid ≥97.0%
3-Amino-2-naphthoic acid
Formel:
C₁₁H₉NO₂ MW: 187,2 g/mol Kokpunkt: 383 °C (1013 hPa) Smältpunkt: 212°C |
MDL Number:
MFCD00004115 CAS nummer: 5959-52-4 EINECS: 227-726-2 Merck Index: 14,00452 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...