1,4-Dibutylbenzene ≥98.0%
Leverantör: TCI EUROPE
D2781-25ML
D2781-5ML
TCIAD2781-25MLEA
4450
SEK
TCIAD2781-25ML
TCIAD2781-5ML
1,4-Dibutylbenzene ≥98.0%
1,4-Dibutylbenzene
Formel:
C₁₄H₂₂ MW: 190,33 g/mol |
MDL Number:
MFCD01076591 CAS nummer: 1571-86-4 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Appearance | Colorless to Almost colorless clear liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...