Methyl-5-chloro-2-nitrobenzoate ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 5-Chloro-2-nitrobenzoic acid methyl ester, Methyl 5-chloro-2-nitrobenzoate, Methyl 5-chloro-2-nitrobenzenecarboxylate
M1996-25G M1996-5G
TCIAM1996-25GEA 630 SEK
TCIAM1996-25G TCIAM1996-5G
Methyl-5-chloro-2-nitrobenzoate ≥98.0%
Methyl-5-chloro-2-nitrobenzoate
Formel: C₈H₆ClNO₄
MW: 215,59 g/mol
Kokpunkt: 306 °C (1013 hPa)
Smältpunkt: 48,5 °C
Densitet: 1,426 g/cm³ (20 °C)
MDL Number: MFCD00272139
CAS nummer: 51282-49-6
EINECS: 257-107-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9c0370f3838967c6ba5476b51812a38940e6184&vid=e9c0370f3838967c6ba5476b51812a38940e6184&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C