1,2,4,5-Tetracyanobenzene ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Fara

Synonyms: Pyromelliticnitrile, 1,2,4,5-Benzenetetranitrile, Benzene-1,2,4,5-tetracarbonitrile, 1,2,4,5-Benzenetetracarbonitrile, Benzene-1,2,4,5-tetracarbonitrile, Pyromellitotetranitrile

T0988-1G T0988-5G
TCIAT0988-1GEA 855 SEK
TCIAT0988-1G TCIAT0988-5G
1,2,4,5-Tetracyanobenzene ≥98.0% (by HPLC, total nitrogen)
1,2,4,5-Tetracyanobenzene
Formel: C₁₀H₂N₄
MW: 178,15 g/mol
Kokpunkt: 374 °C (1013 hPa)
Smältpunkt: 265°C
Densitet: 1,372 g/cm³ (20 °C)
MDL Number: MFCD00043481
CAS nummer: 712-74-3
UN: 3439
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec91df520bad43e553c6a517bca33adf64c8ce93&vid=ec91df520bad43e553c6a517bca33adf64c8ce93&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR