Riboflavin, Millipore®
Leverantör: Merck
Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G
M-Clarity™ quality level = MQ100
1.07609.0010 1.07609.0100
1.07609.0010EA 665 SEK
1.07609.0010 1.07609.0100
Riboflavin, Millipore®
Riboflavin
Formel: C₁₇H₂₀N₄O₆
MW: 376,37 g/mol
Smältpunkt: 280 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005022
CAS nummer: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367