Lauroyl chloride ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Dodecanoyl chloride

D0972-25ML D0972-500ML
TCIAD0972-25MLEA 230 SEK
TCIAD0972-25ML TCIAD0972-500ML
Lauroyl chloride ≥98.0%
Lauroyl chloride
Formel: C₁₂H₂₃ClO
MW: 218,77 g/mol
Kokpunkt: 145°C
Smältpunkt: –12 °C
Densitet: 0,922 g/cm³ (20 °C)
Flampunkt: 132°C
MDL Number: MFCD00000740
CAS nummer: 112-16-3
EINECS: 203-941-7
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eed740ddd73e993eee1af90520f6b9d4782579bc&vid=eed740ddd73e993eee1af90520f6b9d4782579bc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR