Decanoyl chloride ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: Decanoylchloride

D0027-25ML D0027-500ML D0027-100ML
TCIAD0027-25MLEA 255 SEK
TCIAD0027-25ML TCIAD0027-500ML TCIAD0027-100ML
Decanoyl chloride ≥98.0% (by GC, titration analysis)
Decanoyl chloride
Formel: C₁₀H₁₉ClO
MW: 190,71 g/mol
Kokpunkt: 230°C
Smältpunkt: –34,5 °C
Densitet: 0,919 g/cm³ (20 °C)
Flampunkt: 106°C
MDL Number: MFCD00000771
CAS nummer: 112-13-0
EINECS: 203-938-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=955c7f44338c42d05771586628c651992bae820e&vid=955c7f44338c42d05771586628c651992bae820e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR