Pentylene di(acetate)

Leverantör: TCI EUROPE

Synonyms: 1,5-Diacetoxypentane, Pentane-1,5-diyl diacetate, Pentylene glycol diacetate, 1,5-Pentanediol diacetate

P1238-25G P1238-500G
TCIAP1238-25GEA 295 SEK
TCIAP1238-25G TCIAP1238-500G
Pentylene di(acetate)
Pentylene di(acetate)
Formel: C₉H₁₆O₄
MW: 188,22 g/mol
Kokpunkt: 108°C
Smältpunkt: 2 °C
Densitet: 1,033 g/cm³ (25 °C)
MDL Number: MFCD00014985
CAS nummer: 6963-44-6
EINECS: 230-161-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b19b07c9f2418603002675d20c01d5a128168454&vid=b19b07c9f2418603002675d20c01d5a128168454&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR