Etylendi(acetat) ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Ethane-1,2-diyl diacetate, Ethylene glycol diacetate, Ethylene Glycol Diacetate, 1,2-Diacetoxyethane

E0109-25ML E0109-500ML
TCIAE0109-25MLEA 245 SEK
TCIAE0109-25ML TCIAE0109-500ML
Etylendi(acetat) ≥97.0%
Etylendi(acetat)
Formel: C₆H₁₀O₄
MW: 146,14 g/mol
Kokpunkt: 190°C
Smältpunkt: –31 °C
Densitet: 1,104 g/cm³ (20 °C)
Flampunkt: 96°C
Storage Temperature: Omgivande
MDL Number: MFCD00008718
CAS nummer: 111-55-7
EINECS: 203-881-1
Merck Index: 14,03799
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2db99fca7896036a0df989f24f27f2e829565eb6&vid=2db99fca7896036a0df989f24f27f2e829565eb6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR