β-Alanin ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
A0180-25G
A0180-500G
TCIAA0180-25GEA
230
SEK
TCIAA0180-25G
TCIAA0180-500G
β-Alanin ≥99.0%
β-Alanin
Formel:
C₃H₇NO₂ MW: 89,09 g/mol Kokpunkt: 201 °C (1013 hPa) Smältpunkt: 94 °C Densitet: 1,44 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00008200 CAS nummer: 107-95-9 EINECS: 203-536-5 Merck Index: 14,00205 |
Specifikation testresultat
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...