2-Amino-5-bromobenzonitrile ≥98.0%

Leverantör: TCI
Fara

A1960-1G A1960-5G
TCIAA1960-1GEA 455 SEK
TCIAA1960-1G TCIAA1960-5G
2-Amino-5-bromobenzonitrile ≥98.0%
2-Amino-5-bromobenzonitrile
Formel: C₇H₅BrN₂
MDL Number: MFCD00158946
CAS nummer: 39263-32-6
EINECS: 254-387-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62020dc10b1a6554b2c858318cfd4e6dd33d3ad3&vid=62020dc10b1a6554b2c858318cfd4e6dd33d3ad3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 94.0 to 98.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR