5-Bromo-2,3-diaminopyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine
D2916-25G
D2916-5G
TCIAD2916-25GEA
1150
SEK
TCIAD2916-25G
TCIAD2916-5G
5-Bromo-2,3-diaminopyridine ≥98.0% (by GC, titration analysis)
5-Bromo-2,3-diaminopyridine
Formel:
C₅H₆BrN₃ MW: 188,03 g/mol Kokpunkt: 321 °C (1013 hPa) |
MDL Number:
MFCD00460094 CAS nummer: 38875-53-5 EINECS: 254-172-9 ADR: 6.1,III |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...