2-Chloro-5-(chloromethyl)pyridine ≥98.0%

Leverantör: TCI EUROPE
Fara

C1628-25G C1628-5G
TCIAC1628-25GEA 5475 SEK
TCIAC1628-25G TCIAC1628-5G
2-Chloro-5-(chloromethyl)pyridine ≥98.0%
2-Chloro-5-(chloromethyl)pyridine
Formel: C₆H₅Cl₂N
MW: 162,02 g/mol
Smältpunkt: 37 °C
MDL Number: MFCD00125366
CAS nummer: 70258-18-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81daab53c716028952a3a770f421014236368305&vid=81daab53c716028952a3a770f421014236368305&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 41.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR