2-Amino-3-bromo-5-nitrobenzonitrile ≥98.0% (by GC, total nitrogen)
Leverantör: TCI EUROPE
Varning
A1553-25G A1553-5G
TCIAA1553-25GEA 3675 SEK
TCIAA1553-25G TCIAA1553-5G
2-Amino-3-bromo-5-nitrobenzonitrile ≥98.0% (by GC, total nitrogen)
2-Amino-3-bromo-5-nitrobenzonitrile
Formel: C₇H₄BrN₃O₂
MW: 242,03 g/mol
Smältpunkt: 233…235 °C
Densitet: 1,949 g/cm³ (-123 °C)
MDL Number: MFCD00054185
CAS nummer: 17601-94-4
EINECS: 241-574-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17baa8a863818ce9d205a79cba587cfbff3d74aa&vid=17baa8a863818ce9d205a79cba587cfbff3d74aa&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 189.0 to 193.0 deg-C
Solubility in Pyridine almost transparency