D-Alanine ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
(R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
A0177-25G
A0177-5G
TCIAA0177-25GEA
910
SEK
TCIAA0177-25G
TCIAA0177-5G
D-Alanine ≥98.0%
D-Alanine
Formel:
C₃H₇NO₂ MW: 89,09 g/mol Kokpunkt: 189 °C (1013 hPa) Smältpunkt: 79 °C |
MDL Number:
MFCD00008077 CAS nummer: 338-69-2 EINECS: 206-418-1 Merck Index: 13,00203 |
Specifikation testresultat
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -13.0 to -15.0 deg (C=2, 6mol/L HCl) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...