trans-Anethole ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
P0494-25G
P0494-500G
P0494-100G
TCIAP0494-25GEA
230
SEK
TCIAP0494-25G
TCIAP0494-500G
TCIAP0494-100G
trans-Anethole ≥98.0%
trans-Anethole
Formel:
C₁₀H₁₂O MW: 148,2 g/mol Kokpunkt: 235°C Smältpunkt: 21…23 °C Densitet: 0,9883 g/cm³ (20 °C) Flampunkt: 92°C |
MDL Number:
MFCD00009284 CAS nummer: 4180-23-8 EINECS: 224-052-0 Merck Index: 14,00643 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...