Fumaronitrile ≥98.0%

Leverantör: TCI
Fara

F0072-25G F0072-5G
TCIAF0072-25GEA 1875 SEK
TCIAF0072-25G TCIAF0072-5G
Fumaronitrile ≥98.0%
Fumaronitrile
Formel: C₄H₂N₂
Kokpunkt: 186°C
Smältpunkt: 96 °C
Densitet: 1,21 g/cm³ (20 °C)
MDL Number: MFCD00001928
CAS nummer: 764-42-1
EINECS: 212-122-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00549a6762d5340e87c80067e2888a420eedbf38&vid=00549a6762d5340e87c80067e2888a420eedbf38&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR