Allyl hexanoate ≥98.0%
Leverantör: TCI
|
Fara
|
A2179-25ML
A2179-500ML
TCIAA2179-25MLEA
230
SEK
TCIAA2179-25ML
TCIAA2179-500ML
Allyl hexanoate ≥98.0%
Allyl hexanoate
Formel:
C₉H₁₆O₂ Kokpunkt: 188°C Densitet: 0,8886 g/cm³ (20 °C) Flampunkt: 66°C |
MDL Number:
MFCD00038339 CAS nummer: 123-68-2 EINECS: 204-642-4 UN: 2810 ADR: 6.1,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...