Xylitol ≥98.0%
Leverantör: TCI EUROPE
Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
X0018-25G X0018-500G
TCIAX0018-25GEA 215 SEK
TCIAX0018-25G TCIAX0018-500G
Xylitol ≥98.0%
Xylitol
Formel: C₅H₁₂O₅
MW: 152,15 g/mol
Kokpunkt: 495 °C (1013 hPa)
Smältpunkt: 94 °C
Densitet: 1,52 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064292
CAS nummer: 87-99-0
EINECS: 201-788-0
Merck Index: 14,10085
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d831969c9a136fe5831a70aa90976e72eee0de9f&vid=d831969c9a136fe5831a70aa90976e72eee0de9f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Periodic acid Method) min. 98.0 %
Solubility in Water almost transparency