ε-Hexalactone ≥99.0%
Leverantör: TCI EUROPE
Fara
Synonyms: ε-Caprolactone, Oxepan-2-one, Hexan-6-olide
C0702-25ML C0702-500ML
TCIAC0702-25MLEA 230 SEK
TCIAC0702-25ML TCIAC0702-500ML
ε-Hexalactone ≥99.0%
ε-Hexalactone
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 140°C
Smältpunkt: -1°C
Densitet: 1,0705 g/cm³ (20 °C)
Flampunkt: 115°C
MDL Number: MFCD00003267
CAS nummer: 502-44-3
EINECS: 207-938-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=458ebffc6679b26c4c324d3d9150b49f28fb36e8&vid=458ebffc6679b26c4c324d3d9150b49f28fb36e8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Solubility in EtOH almost transparency