Piperidin-4-ol ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
H0413-25G H0413-100G
TCIAH0413-25GEA 725 SEK
TCIAH0413-25G TCIAH0413-100G
Piperidin-4-ol ≥98.0% (by GC, titration analysis)
Piperidin-4-ol
Formel: C₅H₁₁NO
Kokpunkt: 222 °C (1013 hPa)
Smältpunkt: 86…89 °C
Flampunkt: ≤ 100 °C
MDL Number: MFCD00005999
CAS nummer: 5382-16-1
EINECS: 226-373-1
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c79926b76a67dbc605fa4bc3cbf7496f62d7b2a&vid=6c79926b76a67dbc605fa4bc3cbf7496f62d7b2a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 85.0 to 91.0 deg-C
Solubility in Water almost transparency