Piperidin-3-ol ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Piperidinol, 3-Hydroxypiperidine
H0412-25G
H0412-5G
TCIAH0412-25GEA
1125
SEK
TCIAH0412-25G
TCIAH0412-5G
Piperidin-3-ol ≥98.0%
Piperidin-3-ol
Formel:
C₅H₁₁NO MW: 101,15 g/mol Kokpunkt: 113 - 116 °C (1013 hPa) Smältpunkt: 56…59 °C |
MDL Number:
MFCD00014591 CAS nummer: 6859-99-0 EINECS: 229-957-4 UN: 1759 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...