(±)-β-Citronellol ≥95.0%

Leverantör: TCI
Varning

C0370-25ML C0370-500ML C0370-100ML
TCIAC0370-25MLEA 215 SEK
TCIAC0370-25ML TCIAC0370-500ML TCIAC0370-100ML
(±)-β-Citronellol ≥95.0%
(±)-β-Citronellol
Formel: C₁₀H₂₀O
Kokpunkt: 220°C
Densitet: 0,86 g/cm³ (20 °C)
Flampunkt: 102°C
MDL Number: MFCD00002935
CAS nummer: 106-22-9
EINECS: 203-375-0
ADR: 9,III
Merck Index: 14,02330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b72c335a4fb2092662eef87f98cc5d2cf9afd939&vid=b72c335a4fb2092662eef87f98cc5d2cf9afd939&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR