Nitrilotriättiksyra ≥98.0%

Leverantör: TCI
Varning

Synonyms: NTA, 2,2',2''-Nitrilotriacetic acid

N0098-25G N0098-500G
TCIAN0098-25GEA 215 SEK
TCIAN0098-25G TCIAN0098-500G
Nitrilotriättiksyra ≥98.0%
Nitrilotriättiksyra
Formel: C₆H₉NO₆
MW: 191.14 g/mol
Kokpunkt: 498
Smältpunkt: 242
Storage Temperature: Omgivande
MDL Number: MFCD00004287
CAS nummer: 139-13-9
EINECS: 205-355-7
UN: 3077
ADR: 9 III
Merck Index: 14,06579
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f625a3fdac71c8aa4cebcf3936badbc4d6ade19c&vid=f625a3fdac71c8aa4cebcf3936badbc4d6ade19c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency
Ignition residue (Sulfate) max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR