Diklorfen

Leverantör: TCI EUROPE
Varning

Synonyms: 2,2'-Methylenebis(4-chlorophenol), Bis(5-chloro-2-hydroxyphenyl)methane

M0213-25G M0213-500G
TCIAM0213-25GEA 215 SEK
TCIAM0213-25G TCIAM0213-500G
Diklorfen
Diklorfen
Formel: C₁₃H₁₀Cl₂O₂
MW: 269,13 g/mol
Kokpunkt: 389 °C (1013 hPa)
Smältpunkt: 177…178 °C
Flampunkt: ≥ 100 °C
MDL Number: MFCD00002322
CAS nummer: 97-23-4
EINECS: 202-567-1
UN: 3077
ADR: 9,III
Merck Index: 14,03071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b0c97e7d2ca2ec7a9f51eba4281e53e7fd53ad6&vid=4b0c97e7d2ca2ec7a9f51eba4281e53e7fd53ad6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 85.0 area%
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR