3,5-Dimetyl-4-klorfenol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Klor-3,5-xylenol, Klorxylenol

C0161-25G C0161-500G
TCIAC0161-25GEA 245 SEK
TCIAC0161-25G TCIAC0161-500G
3,5-Dimetyl-4-klorfenol ≥98.0%
3,5-Dimetyl-4-klorfenol
Formel: C₈H₉ClO
MW: 156,61 g/mol
Kokpunkt: 246°C
Smältpunkt: 114…116 °C
Densitet: 1,364 g/cm³ (20 °C)
Flampunkt: 132 °C
MDL Number: MFCD00002324
CAS nummer: 88-04-0
EINECS: 201-793-8
UN: 3077
ADR: 9,III
Merck Index: 14,02176
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4153c9be3d6a0c27f29657b0665ab6fdc4140fe2&vid=4153c9be3d6a0c27f29657b0665ab6fdc4140fe2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 114.0 to 117.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR