5-Bromo-2-methoxybenzyl alcohol ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
4-Bromo-2-(hydroxymethyl)anisole
B3369-25G
B3369-5G
TCIAB3369-25GEA
3075
SEK
TCIAB3369-25G
TCIAB3369-5G
5-Bromo-2-methoxybenzyl alcohol ≥98.0%
5-Bromo-2-methoxybenzyl alcohol
Formel:
C₈H₉BrO₂ MW: 217,06 g/mol |
MDL Number:
MFCD00004630 CAS nummer: 80866-82-6 EINECS: 279-586-7 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...