16-Hydroxypalmitic acid ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 16-Hydroxyhexadecylic acid, 16-Hydroxyhexadecanoic acid
H0675-1G H0675-5G
TCIAH0675-1GEA 1025 SEK
TCIAH0675-1G TCIAH0675-5G
16-Hydroxypalmitic acid ≥98.0%
16-Hydroxypalmitic acid
Formel: C₁₆H₃₂O₃
MW: 272,43 g/mol
Kokpunkt: 402 °C (1013 hPa)
Smältpunkt: 95 °C
MDL Number: MFCD00002750
CAS nummer: 506-13-8
EINECS: 208-028-7
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C