Pimelonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Heptanedinitrile, 1,5-Dicyanopentane, Pentamethylene dicyanide

P0049-25ML P0049-500ML
TCIAP0049-25MLEA 605 SEK
TCIAP0049-25ML TCIAP0049-500ML
Pimelonitrile ≥98.0%
Pimelonitrile
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
Kokpunkt: 175°C
Smältpunkt: –31 °C
Densitet: 0,95 g/cm³ (20 °C)
Flampunkt: ≥ 112 °C
MDL Number: MFCD00001980
CAS nummer: 646-20-8
EINECS: 211-469-8
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9ccff2d1dc318018dcf1f5252a178fb9df3d2cc&vid=f9ccff2d1dc318018dcf1f5252a178fb9df3d2cc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR