6-Nitro-2,4-xylenol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2,4-Dimethyl-6-nitrophenol, 4-Hydroxy-5-nitro-3-xylene

D2273-25G D2273-5G
TCIAD2273-25GEA 2850 SEK
TCIAD2273-25G TCIAD2273-5G
6-Nitro-2,4-xylenol ≥98.0% (by GC, titration analysis)
6-Nitro-2,4-xylenol
Formel: C₈H₉NO₃
MW: 167,16 g/mol
MDL Number: MFCD00191633
CAS nummer: 14452-34-7
EINECS: 238-436-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5f845da7c70b5ce496d8eccab88da1ac4dc57e4&vid=e5f845da7c70b5ce496d8eccab88da1ac4dc57e4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR