N,N'-Diethylethylenediamine ≥95.0%

Leverantör: TCI EUROPE
Fara

D0287-25ML D0287-5ML
TCIAD0287-25MLEA 1425 SEK
TCIAD0287-25ML TCIAD0287-5ML
N,N'-Diethylethylenediamine ≥95.0%
N,N'-Diethylethylenediamine
Formel: C₆H₁₆N₂
MW: 116,21 g/mol
Kokpunkt: 150°C
Smältpunkt: –11 °C
Densitet: 0,811 g/cm³ (20 °C)
Flampunkt: 33°C
MDL Number: MFCD00009033
CAS nummer: 111-74-0
EINECS: 203-902-4
UN: 2685
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4be3f60b1a219182a15ae063a47ba174b574bab&vid=f4be3f60b1a219182a15ae063a47ba174b574bab&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR