Trimethylsulphonium iodide ≥98.0%
Leverantör: TCI
|
Varning
|
T1056-25G
T1056-500G
TCIAT1056-25GEA
335
SEK
TCIAT1056-25G
TCIAT1056-500G
Trimethylsulphonium iodide ≥98.0%
Trimethylsulphonium iodide
Formel:
C₃H₉IS Smältpunkt: 215…220 °C Densitet: 1,922 g/cm³ (20 °C) |
MDL Number:
MFCD00011632 CAS nummer: 2181-42-2 EINECS: 218-555-4 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Potassium iodate Method) | min. 98.0 % |
Melting point | 200.0 to 206.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...