Methyl crotonate ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Methyl trans-2-butenoate, 2-Butenoic acid methyl ester, trans-Methyl crotonate, trans-Crotonic acid methyl ester, Crotonic acid methyl ester, trans-2-Butenoic acid methyl ester
C0419-25ML
C0419-500ML
C0419-100ML
TCIAC0419-25MLEA
390
SEK
TCIAC0419-25ML
TCIAC0419-500ML
TCIAC0419-100ML
Methyl crotonate ≥98.0%
Methyl crotonate
Formel:
C₅H₈O₂ Kokpunkt: 120°C Densitet: 0,95 g/cm³ (20 °C) Flampunkt: 29°C |
MDL Number:
MFCD00009287 CAS nummer: 623-43-8 EINECS: 210-793-7 UN: 3272 ADR: 3,III Merck Index: 14,02597 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...