Thiazole ≥98.0%
Leverantör: TCI
|
Varning
|
T0185-25G
T0185-5G
TCIAT0185-25GEA
2125
SEK
TCIAT0185-25G
TCIAT0185-5G
Thiazole ≥98.0%
Thiazole
Formel:
C₃H₃NS Kokpunkt: 118°C Smältpunkt: –34 °C Densitet: 1,2 g/cm³ (20 °C) Flampunkt: 26°C |
MDL Number:
MFCD00005315 CAS nummer: 288-47-1 EINECS: 206-021-3 UN: 1993 ADR: 3,III Merck Index: 14,09307 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...