Tetrafluorophthalic anhydride ≥95.0%

Leverantör: TCI
Varning

T2381-1G T2381-5G
TCIAT2381-1GEA 365 SEK
TCIAT2381-1G TCIAT2381-5G
Tetrafluorophthalic anhydride ≥95.0%
Tetrafluorophthalic anhydride
Formel: C₈F₄O₃
MDL Number: MFCD00039697
CAS nummer: 652-12-0
EINECS: 211-485-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84d639ece5801bd71de221e145bb3646aecd2bd3&vid=84d639ece5801bd71de221e145bb3646aecd2bd3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Ethanolysis Method) min. 95.0 %
Melting point 92.0 to 96.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR