Tetrabromophthalic anhydride ≥95.0%

Leverantör: TCI
Varning

T0046-25G T0046-100G
TCIAT0046-25GEA 215 SEK
TCIAT0046-25G TCIAT0046-100G
Tetrabromophthalic anhydride ≥95.0%
Formel: C₈Br₄O₃
Kokpunkt: 477 °C (1013 hPa)
Smältpunkt: 280°C
Densitet: 2,93 g/cm³ (20 °C)
MDL Number: MFCD00005919
CAS nummer: 632-79-1
EINECS: 211-185-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26d21f53485cd599e793df00fae1a327486aa9cc&vid=26d21f53485cd599e793df00fae1a327486aa9cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR