Methyl propiolate ≥98.0%
Leverantör: TCI
|
Fara
|
P0528-25ML
P0528-5ML
TCIAP0528-25MLEA
1750
SEK
TCIAP0528-25ML
TCIAP0528-5ML
Methyl propiolate ≥98.0%
Methyl propiolate
Formel:
C₄H₄O₂ Kokpunkt: 102°C Densitet: 1,196 g/cm³ (20 °C) Flampunkt: 10°C |
MDL Number:
MFCD00008572 CAS nummer: 922-67-8 EINECS: 213-083-5 UN: 3272 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...