Ethyl propiolate ≥98.0%

Leverantör: TCI
Varning

Synonyms: Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester

P0529-25ML P0529-5ML
TCIAP0529-25MLEA 1525 SEK
TCIAP0529-25ML TCIAP0529-5ML
Ethyl propiolate ≥98.0%
Ethyl propiolate
Formel: C₅H₆O₂
Kokpunkt: 119°C
Smältpunkt: 9 °C
Densitet: 0,9788 g/cm³ (4 °C)
Flampunkt: 25°C
MDL Number: MFCD00009184
CAS nummer: 623-47-2
EINECS: 210-795-8
UN: 3272
ADR: 3,III
Merck Index: 14,03846
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01895050f409b64d03b793aa47a09af69812152b&vid=01895050f409b64d03b793aa47a09af69812152b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR