N,N,N-Trimetylfenylammoniumklorid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Trimethylphenylammonium chloride, Phenyltrimethylammonium chloride
P0244-25G P0244-500G P0244-100G
TCIAP0244-25GEA 215 SEK
TCIAP0244-25G TCIAP0244-500G TCIAP0244-100G
N,N,N-Trimetylfenylammoniumklorid ≥98.0% (by HPLC, titration analysis)
N,N,N-Trimetylfenylammoniumklorid
Formel: C₉H₁₄ClN
MW: 171,67 g/mol
Smältpunkt: 246…247 °C
MDL Number: MFCD00011790
CAS nummer: 138-24-9
EINECS: 205-319-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b43b550cb24c08a226f710fd2b200983978051&vid=11b43b550cb24c08a226f710fd2b200983978051&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency