N-Metylformamid ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Formmethylamide, Formic acid methylamide

F0059-25G F0059-500G
TCIAF0059-25GEA 230 SEK
TCIAF0059-25G TCIAF0059-500G
N-Metylformamid ≥99.0%
N-Metylformamid
Formel: C₂H₅NO
MW: 59,07 g/mol
Kokpunkt: 183°C
Smältpunkt: -5°C
Densitet: 1,011 g/cm³ (20 °C)
Flampunkt: 98°C
MDL Number: MFCD00003280
CAS nummer: 123-39-7
EINECS: 204-624-6
Merck Index: 14,06076
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a560dc532404df3e63cc8a24531b916a0f4f7ed&vid=5a560dc532404df3e63cc8a24531b916a0f4f7ed&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR