Dimethyl sulphone ≥99.0%

Leverantör: TCI

M1239-25G M1239-500G
TCIAM1239-25GEA 215 SEK
TCIAM1239-25G TCIAM1239-500G
Dimethyl sulphone ≥99.0%
Dimethyl sulphone
Formel: C₂H₆O₂S
Kokpunkt: 238°C
Smältpunkt: 107…110 °C
Densitet: 1,17 g/cm³ (110 °C)
MDL Number: MFCD00007566
CAS nummer: 67-71-0
EINECS: 200-665-9
Merck Index: 14,03258
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=398fd01b744f4bfee84672b994bc679d2b8e7eb1&vid=398fd01b744f4bfee84672b994bc679d2b8e7eb1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR