Methyl dichloroacetate ≥99.0%

Leverantör: TCI EUROPE
Fara

D0309-25G D0309-500G
TCIAD0309-25GEA 215 SEK
TCIAD0309-25G TCIAD0309-500G
Methyl dichloroacetate ≥99.0%
Methyl dichloroacetate
Formel: C₃H₄Cl₂O₂
MW: 142,97 g/mol
Kokpunkt: 143°C
Smältpunkt: –52 °C
Densitet: 1,3774 g/cm³ (20 °C)
Flampunkt: 72°C
MDL Number: MFCD00000843
CAS nummer: 116-54-1
EINECS: 204-146-8
UN: 2299
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=047457e9ebee77c7595a73cfecb18a77bd2ebdcb&vid=047457e9ebee77c7595a73cfecb18a77bd2ebdcb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR