Dibenzyl diselenide ≥95.0%

Leverantör: TCI
Fara

D2717-5G
TCIAD2717-5GEA 1425 SEK
TCIAD2717-5G
Dibenzyl diselenide ≥95.0%
Dibenzyl diselenide
Formel: C₁₄H₁₄Se₂
MW: 340.19 g/mol
MDL Number: MFCD00004767
CAS nummer: 1482-82-2
UN: 3283
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64656e246ab11ff98954260d8dbb283ea2d42578&vid=64656e246ab11ff98954260d8dbb283ea2d42578&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (Iodometric Titration) min. 95.0 %
Melting point 91.0 to 95.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR