N-Phenylmaleimide ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
P0900-25G
P0900-500G
P0900-100G
TCIAP0900-25GEA
230
SEK
TCIAP0900-25G
TCIAP0900-500G
TCIAP0900-100G
N-Phenylmaleimide ≥98.0%
N-Phenylmaleimide
Formel:
C₁₀H₇NO₂ MW: 173,17 g/mol Kokpunkt: 181°C Smältpunkt: 84…86 °C |
MDL Number:
MFCD00005502 CAS nummer: 941-69-5 EINECS: 213-382-0 UN: 2811 ADR: 6.1,III Merck Index: 14,07299 |
Specifikation testresultat
Appearance | Light yellow to Yellow flake |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...