7-Nitro-1H-indazole ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole
N0827-1G N0827-5G
TCIAN0827-1GEA 815 SEK
TCIAN0827-1G TCIAN0827-5G
7-Nitro-1H-indazole ≥97.0%
7-Nitro-1H-indazole
Formel: C₇H₅N₃O₂
MW: 163,14 g/mol
Smältpunkt: 190 °C
Densitet: 1,56 g/cm³ (293 °C)
MDL Number: MFCD00022789
CAS nummer: 2942-42-9
EINECS: 220-934-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d186197eaafa47f2d64f48c23b9038d68ee89a0a&vid=d186197eaafa47f2d64f48c23b9038d68ee89a0a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 184.0 to 188.0 deg-C