Indole-5-carbaldehyd ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Indole-5-carboxaldehyde, 1H-Indole-5-carboxaldehyde
I0674-25G
I0674-5G
TCIAI0674-25GEA
6125
SEK
TCIAI0674-25G
TCIAI0674-5G
Indole-5-carbaldehyd ≥98.0%
Indole-5-carbaldehyd
Formel:
C₉H₇NO MW: 145,16 g/mol |
MDL Number:
MFCD02093664 CAS nummer: 1196-69-6 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...