5-Hexenoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 5-Hexenoicacid

H0875-25G H0875-5G
TCIAH0875-25GEA 6150 SEK
TCIAH0875-25G TCIAH0875-5G
5-Hexenoic acid ≥98.0% (by GC, titration analysis)
5-Hexenoic acid
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 203°C
Smältpunkt: –37 °C
Densitet: 0,961 g/cm³ (20 °C)
Flampunkt: 104°C
MDL Number: MFCD00046558
CAS nummer: 1577-22-6
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29443ea1ad51804153685c48606f9cdbaf4379a9&vid=29443ea1ad51804153685c48606f9cdbaf4379a9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR