Fluorene-9-carboxylic acid ≥97.0%
Leverantör: TCI EUROPE
Synonyms: 9H-Fluorene-9-carboxylic acid
F0022-25G F0022-5G
TCIAF0022-25GEA 1800 SEK
TCIAF0022-25G TCIAF0022-5G
Fluorene-9-carboxylic acid ≥97.0%
Fluorene-9-carboxylic acid
Formel: C₁₄H₁₀O₂
MW: 210,23 g/mol
Kokpunkt: 365 °C (1013 hPa)
Smältpunkt: 226…229 °C
Densitet: 1,333 g/cm³ (20 °C)
MDL Number: MFCD00001136
CAS nummer: 1989-33-9
EINECS: 217-866-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8a7058c3fd9a9cd56d9ef9eada75aa73e8acc4c&vid=e8a7058c3fd9a9cd56d9ef9eada75aa73e8acc4c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 94.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 229.0 to 234.0 deg-C