5-Aminouracil ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Aminouracil, 5-Aminopyrimidine-2,4(1H,3H)-dione
A0898-25G A0898-5G
TCIAA0898-25GEA 3325 SEK
TCIAA0898-25G TCIAA0898-5G
5-Aminouracil ≥98.0%
5-Aminouracil
Formel: C₄H₅N₃O₂
MW: 127,1 g/mol
Kokpunkt: 377 °C (1013 hPa)
Smältpunkt: 300 °C
MDL Number: MFCD00006025
CAS nummer: 932-52-5
EINECS: 213-252-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3993215331730d5d06b457df3637101e23fdab&vid=db3993215331730d5d06b457df3637101e23fdab&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %